siRNA Details
| Gene Symbol: Smad5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021692.1 | NP_067724.1 | 1 | 306 | ||
| XM_006253586.2 | XP_006253648.1 | 4 | 771 | |||
| XM_006253587.2 | XP_006253649.1 | 3 | 689 | |||
| XM_006253588.2 | XP_006253650.1 | 4 | 1013 | |||
| XM_008771446.1 | XP_008769668.1 | 3 | 2349 | |||
| XM_008771447.1 | XP_008769669.1 | 4 | 1531 | |||
| XM_008771448.1 | XP_008769670.1 | 4 | 1706 | |||
| GenBank mRNA | AB010955.1 | - | 1 | 306 | ||
| BC070905.1 | - | 4 | 701 | |||
| BC161849.1 | - | 2 | 493 | 9099068 | ||

Smad5
Genomic Map