siRNA Details
| Gene Symbol: Synpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021695.2 | NP_067727.2 | 2 | 1806 | ||
| GenBank mRNA | AB013130.1 | - | 1 | 1332 | ||
| AM980944.1 | - | 1276 | ||||
| DY313339.1 | - | 1 | 424 | |||
| EV779156.1 | - | 1 | 185 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Synpo| Gene Symbol: Synpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021695.2 | NP_067727.2 | 2 | 1806 | ||
| GenBank mRNA | AB013130.1 | - | 1 | 1332 | ||
| AM980944.1 | - | 1276 | ||||
| DY313339.1 | - | 1 | 424 | |||
| EV779156.1 | - | 1 | 185 | |||