siRNA Details
| Gene Symbol: Serpinb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021696.2 | NP_067728.1 | 1 | 51 | ||
| XM_006249639.2 | XP_006249701.1 | 2 | 237 | |||
| XM_008769495.1 | XP_008767717.1 | 2 | 134 | |||
| GenBank mRNA | X64563.1 | - | 1 | 52 | ||

Serpinb2
Genomic Map