siRNA Details
| Gene Symbol: Mark2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021699.1 | NP_067731.1 | 8 | 660 | ||
| XM_006230879.2 | XP_006230941.1 | 8 | 1160 | |||
| XM_006230880.2 | XP_006230942.1 | 8 | 1947 | |||
| XM_006230881.2 | XP_006230943.1 | 8 | 679 | |||
| XM_006230883.2 | XP_006230945.1 | 8 | 1159 | |||
| XM_006230884.2 | XP_006230946.1 | 8 | 1158 | |||
| XM_006230885.2 | XP_006230947.1 | 8 | 1158 | |||
| XM_006230887.2 | XP_006230949.1 | 8 | 1157 | |||
| XM_008760169.1 | XP_008758391.1 | 8 | 1159 | |||
| GenBank mRNA | Z83869.1 | - | 8 | 660 | ||

Mark2
Genomic Map