siRNA Details
| Gene Symbol: Msln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031658.1 | NP_113846.1 | 5, 6 | 475 | ||
| XM_006246021.2 | XP_006246083.1 | 6, 7 | 426 | |||
| XM_006246022.2 | XP_006246084.1 | 6, 7 | 433 | |||
| GenBank mRNA | BC070934.1 | - | 5, 6 | 436 | 7098161 | |
| D87351.1 | - | 5, 6 | 475 | |||

Msln
Genomic Map