siRNA Details
| Gene Symbol: Cep78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021741.2 | NP_068509.2 | 15 | 2184 | ||
| XM_006231136.2 | XP_006231198.1 | 15 | 2245 | |||
| GenBank mRNA | FQ222521.1 | - | 13 | 1995 | ||
| X99330.1 | - | 15 | 1918 | |||

Cep78
Genomic Map