siRNA Details
| Gene Symbol: Nr5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021742.1 | NP_068510.1 | 7 | 1315 | ||
| XM_006249928.2 | XP_006249990.1 | 6 | 1423 | |||
| XM_006249929.2 | XP_006249991.1 | 5 | 1163 | |||
| GenBank mRNA | AB012960.1 | - | 7 | 1315 | ||
| AB012961.1 | - | 6 | 1252 | |||

Nr5a2
Genomic Map