siRNA Details
| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021746.1 | NP_068514.1 | 9, 10 | 956 | ||
| XM_008765781.1 | XP_008764003.1 | 9, 10 | 685 | |||
| GenBank mRNA | X96488.1 | - | 9, 10 | 956 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mapk12| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021746.1 | NP_068514.1 | 9, 10 | 956 | ||
| XM_008765781.1 | XP_008764003.1 | 9, 10 | 685 | |||
| GenBank mRNA | X96488.1 | - | 9, 10 | 956 | ||