siRNA Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021748.1 | NP_068516.1 | 763 | |||
| GenBank mRNA | AF142097.1 | - | 763 | |||
| AF189019.1 | - | 763 | ||||
| FQ211783.1 | - | 802 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nsf| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021748.1 | NP_068516.1 | 763 | |||
| GenBank mRNA | AF142097.1 | - | 763 | |||
| AF189019.1 | - | 763 | ||||
| FQ211783.1 | - | 802 | ||||