siRNA Details
| Gene Symbol: Pga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021753.1 | NP_068521.1 | 3, 4 | 355 | ||
| GenBank mRNA | AJ251687.1 | - | 3, 4 | 355 | ||
| Gene Symbol: LOC100910889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749058.3 | XP_003749106.1 | 3, 4 | 1014 | ||

Pga5
Genomic Map