siRNA Details
| Gene Symbol: Nop58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021754.1 | NP_068522.1 | 11 | 1299 | ||
| XM_006245013.1 | XP_006245075.1 | 10 | 1124 | |||
| GenBank mRNA | AF194371.1 | - | 11 | 1299 | ||
| BC087637.1 | - | 11 | 1332 | 7309062 | ||
| FQ225580.1 | - | 4 | 495 | |||
| FQ230388.1 | - | 4 | 495 | |||

Nop58
Genomic Map