siRNA Details
| Gene Symbol: Lmna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002016.2 | NP_001002016.2 | 8 | 1658 | ||
| GenBank mRNA | BC062018.1 | - | 7 | 1213 | 5621637 | |
| CA338388.1 | - | 4 | 643 | |||
| FM060776.1 | - | 2 | 195 | |||
| X66870.1 | - | 8 | 1617 | |||
| X76297.1 | - | 8 | 1380 | |||
| X99257.1 | - | 8 | 1117 | |||

Lmna
Genomic Map