siRNA Details
| Gene Symbol: Lmna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002016.2 | NP_001002016.2 | 10 | 1847 | ||
| GenBank mRNA | BC062018.1 | - | 9 | 1402 | 5621637 | |
| FM060776.1 | - | 4 | 384 | |||
| X66870.1 | - | 10 | 1806 | |||
| X76297.1 | - | 10 | 1569 | |||
| X99257.1 | - | 10 | 1306 | |||

Lmna
Genomic Map