siRNA Details
| Gene Symbol: Lin7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021758.1 | NP_068526.1 | 4 | 286 | ||
| GenBank mRNA | AF090133.1 | - | 4 | 286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lin7b| Gene Symbol: Lin7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021758.1 | NP_068526.1 | 4 | 286 | ||
| GenBank mRNA | AF090133.1 | - | 4 | 286 | ||