siRNA Details
| Gene Symbol: Col5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021760.1 | NP_068528.1 | 66 | 4936 | ||
| XM_008765979.1 | XP_008764201.1 | 66 | 5001 | |||
| GenBank mRNA | AF272661.1 | - | 66 | 4936 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col5a3| Gene Symbol: Col5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021760.1 | NP_068528.1 | 66 | 4936 | ||
| XM_008765979.1 | XP_008764201.1 | 66 | 5001 | |||
| GenBank mRNA | AF272661.1 | - | 66 | 4936 | ||