siRNA Details
| Gene Symbol: Copb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021765.1 | NP_068533.1 | 269 | |||
| GenBank mRNA | AF002705.1 | - | 269 | |||
| BC064317.1 | - | 289 | ||||
| BC088397.1 | - | 244 | 7190660 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Copb2| Gene Symbol: Copb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021765.1 | NP_068533.1 | 269 | |||
| GenBank mRNA | AF002705.1 | - | 269 | |||
| BC064317.1 | - | 289 | ||||
| BC088397.1 | - | 244 | 7190660 | |||