siRNA Details
Gene Symbol: Gde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032615.2 | NP_116004.2 | 687 | |||
GenBank mRNA | AF212861.1 | - | 632 | |||
BC070897.1 | - | 687 | 7099596 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032615.2 | NP_116004.2 | 687 | |||
GenBank mRNA | AF212861.1 | - | 632 | |||
BC070897.1 | - | 687 | 7099596 |