siRNA Details
| Gene Symbol: Slc18a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031663.2 | NP_113851.1 | 1 | 1455 | ||
| GenBank mRNA | U09211.1 | - | 1 | 1455 | ||
| U09838.1 | - | 1 | 712 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc18a3| Gene Symbol: Slc18a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031663.2 | NP_113851.1 | 1 | 1455 | ||
| GenBank mRNA | U09211.1 | - | 1 | 1455 | ||
| U09838.1 | - | 1 | 712 | |||