siRNA Details
| Gene Symbol: Kcnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021853.1 | NP_068625.1 | 17 | 1639 | ||
| XM_006233697.2 | XP_006233759.1 | 17 | 1947 | |||
| XM_006233698.2 | XP_006233760.1 | 17 | 1947 | |||
| XM_006233699.2 | XP_006233761.1 | 17 | 1947 | |||
| XM_006233700.2 | XP_006233762.1 | 16 | 1905 | |||
| XM_006233701.2 | XP_006233763.1 | 17 | 1947 | |||
| XM_006233702.2 | XP_006233764.1 | 16 | 1905 | |||
| XM_006233703.2 | XP_006233765.1 | 17 | 1947 | |||
| XM_006233704.2 | XP_006233766.1 | 16 | 1827 | |||
| XM_006233705.2 | XP_006233767.1 | 16 | 1824 | |||
| XM_006233706.2 | XP_006233768.1 | 17 | 1947 | |||
| XM_006233707.2 | XP_006233769.1 | 16 | 1822 | |||
| XM_008761624.1 | XP_008759846.1 | 17 | 1937 | |||
| GenBank mRNA | AF089730.1 | - | 17 | 1639 | ||
| AY884213.1 | - | 16 | 1537 | |||

Kcnt1
Genomic Map