siRNA Details
| Gene Symbol: Fgf16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021867.3 | NP_068639.1 | 3 | 447 | ||
| GenBank mRNA | AB002561.1 | - | 3 | 447 | ||
| BC085738.1 | - | 721 | 7189721 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf16| Gene Symbol: Fgf16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021867.3 | NP_068639.1 | 3 | 447 | ||
| GenBank mRNA | AB002561.1 | - | 3 | 447 | ||
| BC085738.1 | - | 721 | 7189721 | |||