siRNA Details
| Gene Symbol: Stx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021869.2 | NP_068641.2 | 4 | 268 | ||
| XM_006227691.2 | XP_006227753.1 | 5 | 1590 | |||
| XM_008758626.1 | XP_008756848.1 | 5 | 1584 | |||
| GenBank mRNA | AF031430.1 | - | 3 | 172 | ||
| BC085737.1 | - | 4 | 268 | 7193349 | ||
| Gene Symbol: LOC100910446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758670.1 | XP_008756892.1 | 2 | 154 | ||

Stx7
Genomic Map