siRNA Details
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031665.2 | NP_113853.1 | 3 | 449 | ||
GenBank mRNA | BC081769.1 | - | 3 | 449 | 7134641 | |
U56815.1 | - | 3 | 425 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031665.2 | NP_113853.1 | 3 | 449 | ||
GenBank mRNA | BC081769.1 | - | 3 | 449 | 7134641 | |
U56815.1 | - | 3 | 425 |