siRNA Details
| Gene Symbol: Mybbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031668.1 | NP_113856.1 | 3 | 211 | ||
| GenBank mRNA | U83590.1 | - | 3 | 211 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mybbp1a| Gene Symbol: Mybbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031668.1 | NP_113856.1 | 3 | 211 | ||
| GenBank mRNA | U83590.1 | - | 3 | 211 | ||