siRNA Details
Gene Symbol: Mybbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031668.1 | NP_113856.1 | 8 | 1096 | ||
GenBank mRNA | U83590.1 | - | 8 | 1096 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mybbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031668.1 | NP_113856.1 | 8 | 1096 | ||
GenBank mRNA | U83590.1 | - | 8 | 1096 |