siRNA Details
| Gene Symbol: Napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031670.2 | NP_113858.1 | 9 | 1253 | ||
| GenBank mRNA | AJ251299.1 | - | 9 | 1225 | ||
| BC078790.1 | - | 9 | 1245 | 7109515 | ||
| FQ225885.1 | - | 9 | 1231 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Napsa| Gene Symbol: Napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031670.2 | NP_113858.1 | 9 | 1253 | ||
| GenBank mRNA | AJ251299.1 | - | 9 | 1225 | ||
| BC078790.1 | - | 9 | 1245 | 7109515 | ||
| FQ225885.1 | - | 9 | 1231 | |||