siRNA Details
| Gene Symbol: Acaca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022193.1 | NP_071529.1 | 46 | 6181 | ||
| GenBank mRNA | EF121986.1 | - | 46 | 6181 | ||
| EF121987.1 | - | 46 | 6181 | |||
| J03808.1 | - | 46 | 6181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acaca| Gene Symbol: Acaca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022193.1 | NP_071529.1 | 46 | 6181 | ||
| GenBank mRNA | EF121986.1 | - | 46 | 6181 | ||
| EF121987.1 | - | 46 | 6181 | |||
| J03808.1 | - | 46 | 6181 | |||