siRNA Details
| Gene Symbol: Clcn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022198.2 | NP_071534.2 | 4 | 426 | ||
| XM_006256828.2 | XP_006256890.1 | 4 | 729 | |||
| GenBank mRNA | BC092209.1 | - | 4 | 426 | 7315063 | |
| FQ210965.1 | - | 4 | 465 | |||
| Z36944.1 | - | 2 | 134 | |||

Clcn4
Genomic Map