siRNA Details
| Gene Symbol: Epha8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764322.1 | XP_008762544.1 | 9 | 1683 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epha8| Gene Symbol: Epha8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764322.1 | XP_008762544.1 | 9 | 1683 | ||