siRNA Details
| Gene Symbol: Exoc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022204.3 | NP_071540.1 | 14 | 1690 | ||
| GenBank mRNA | BC128739.1 | - | 14 | 1693 | 8362342 | |
| FQ217124.1 | - | 13 | 1524 | |||
| U79417.1 | - | 14 | 1462 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exoc5| Gene Symbol: Exoc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022204.3 | NP_071540.1 | 14 | 1690 | ||
| GenBank mRNA | BC128739.1 | - | 14 | 1693 | 8362342 | |
| FQ217124.1 | - | 13 | 1524 | |||
| U79417.1 | - | 14 | 1462 | |||