siRNA Details
Gene Symbol: Pik3r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022213.1 | NP_071549.1 | 6 | 706 | ||
XM_006238714.2 | XP_006238776.1 | 7 | 727 | |||
XM_008763979.1 | XP_008762201.1 | 7 | 929 | |||
GenBank mRNA | D64047.1 | - | 6 | 706 |
