siRNA Details
| Gene Symbol: Tmeff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023020.2 | NP_075409.1 | 3, 4 | 712 | ||
| GenBank mRNA | AJ250730.1 | - | 3, 4 | 509 | ||
| BC129093.1 | - | 3, 4 | 712 | 8363015 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmeff1| Gene Symbol: Tmeff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023020.2 | NP_075409.1 | 3, 4 | 712 | ||
| GenBank mRNA | AJ250730.1 | - | 3, 4 | 509 | ||
| BC129093.1 | - | 3, 4 | 712 | 8363015 | ||