siRNA Details
| Gene Symbol: Hsd17b10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031682.1 | NP_113870.1 | 4 | 447 | ||
| GenBank mRNA | AF049878.1 | - | 4 | 435 | ||
| AF069770.1 | - | 4 | 447 | |||
| BC158587.1 | - | 4 | 441 | 9089087 | ||
| FQ210583.1 | - | 2 | 188 | |||
| FQ220319.1 | - | 2 | 188 | |||
| FQ225948.1 | - | 2 | 188 | |||
| FQ228203.1 | - | 2 | 188 | |||
| FQ231826.1 | - | 2 | 188 | |||
| FQ232016.1 | - | 2 | 188 | |||

Hsd17b10
Genomic Map