siRNA Details
| Gene Symbol: Pde10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022236.1 | NP_071572.1 | 21, 22 | 2557 | ||
| GenBank mRNA | AB027155.1 | - | 21, 22 | 2557 | ||
| AB027156.1 | - | 2892 | ||||
| AY462091.1 | - | 2523 | ||||
| AY462092.1 | - | 2631 | ||||
| AY462093.1 | - | 2669 | ||||
| AY462094.1 | - | 2338 | ||||
| AY462095.1 | - | 2804 | ||||

Pde10a
Genomic Map