siRNA Details
| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022242.2 | NP_071578.2 | 14 | 2501 | ||
| GenBank mRNA | AB046718.2 | - | 14 | 2501 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam129a| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022242.2 | NP_071578.2 | 14 | 2501 | ||
| GenBank mRNA | AB046718.2 | - | 14 | 2501 | ||