siRNA Details
| Gene Symbol: Arhgap17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270692.1 | NP_001257621.1 | 9, 10 | 799 | ||
| NM_001270693.1 | NP_001257622.1 | 9, 10 | 799 | |||
| NM_001270694.1 | NP_001257623.1 | 9, 10 | 799 | |||
| NM_022244.2 | NP_071580.1 | 9, 10 | 799 | |||
| XM_006230194.2 | XP_006230256.1 | 9, 10 | 851 | |||
| XM_008759791.1 | XP_008758013.1 | 9, 10 | 851 | |||
| XM_008759792.1 | XP_008758014.1 | 9, 10 | 852 | |||
| XM_008759793.1 | XP_008758015.1 | 9, 10 | 851 | |||
| XM_008759794.1 | XP_008758016.1 | 9, 10 | 850 | |||
| GenBank mRNA | AB042827.1 | - | 9, 10 | 798 | ||
| AB060556.1 | - | 9, 10 | 798 | |||
| AB060557.1 | - | 9, 10 | 798 | |||
| AB080637.1 | - | 9, 10 | 798 | |||
| BC085736.1 | - | 9, 10 | 800 | 7130218 | ||

Arhgap17
Genomic Map