siRNA Details
| Gene Symbol: Slc29a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031684.2 | NP_113872.1 | 6 | 618 | ||
| XM_006244560.1 | XP_006244622.1 | 6 | 649 | |||
| XM_006244561.2 | XP_006244623.1 | 7 | 882 | |||
| XM_006244562.2 | XP_006244624.1 | 6 | 821 | |||
| XM_006244563.2 | XP_006244625.1 | 6 | 644 | |||
| XM_008766879.1 | XP_008765101.1 | 7 | 707 | |||
| GenBank mRNA | AF015304.1 | - | 3 | 304 | ||
| BC078789.1 | - | 6 | 618 | 7132212 | ||

Slc29a1
Genomic Map