siRNA Details
| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022246.1 | NP_071582.1 | 16 | 2832 | ||
| GenBank mRNA | AF218576.1 | - | 16 | 2832 | ||
| FQ232550.1 | - | 14 | 2356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad50| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022246.1 | NP_071582.1 | 16 | 2832 | ||
| GenBank mRNA | AF218576.1 | - | 16 | 2832 | ||
| FQ232550.1 | - | 14 | 2356 | |||