siRNA Details
Gene Symbol: Pdcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022247.1 | NP_071583.1 | 4 | 717 | ||
GenBank mRNA | BC097322.1 | - | 4 | 711 | 7453703 | |
FQ211647.1 | - | 703 | ||||
FQ223348.1 | - | 2115 | ||||
L15354.1 | - | 4 | 717 | |||
L15355.1 | - | 2 | 590 |
Gene Symbol: LOC690155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761806.1 | XP_008760028.1 | 4 | 977 |
