siRNA Details
| Gene Symbol: Qtrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022250.2 | NP_071586.2 | 3 | 410 | ||
| GenBank mRNA | AB034634.1 | - | 3 | 405 | ||
| BC097321.1 | - | 3 | 410 | 7387393 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qtrt1| Gene Symbol: Qtrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022250.2 | NP_071586.2 | 3 | 410 | ||
| GenBank mRNA | AB034634.1 | - | 3 | 405 | ||
| BC097321.1 | - | 3 | 410 | 7387393 | ||