siRNA Details
| Gene Symbol: Gpr173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022255.1 | NP_071591.1 | 1 | 906 | ||
| GenBank mRNA | AB040804.1 | - | 1 | 906 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr173| Gene Symbol: Gpr173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022255.1 | NP_071591.1 | 1 | 906 | ||
| GenBank mRNA | AB040804.1 | - | 1 | 906 | ||
Set Membership: |
|