siRNA Details
| Gene Symbol: Cd244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022259.1 | NP_071595.1 | 2 | 404 | ||
| XM_006250259.2 | XP_006250321.1 | 2 | 1783 | |||
| GenBank mRNA | AF156988.1 | - | 2 | 464 | ||
| AF156989.1 | - | 2 | 404 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd244| Gene Symbol: Cd244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022259.1 | NP_071595.1 | 2 | 404 | ||
| XM_006250259.2 | XP_006250321.1 | 2 | 1783 | |||
| GenBank mRNA | AF156988.1 | - | 2 | 464 | ||
| AF156989.1 | - | 2 | 404 | |||