siRNA Details
| Gene Symbol: Casp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022260.3 | NP_071596.1 | 2 | 204 | ||
| XM_008760546.1 | XP_008758768.1 | 3 | 797 | |||
| XM_008760547.1 | XP_008758769.1 | 2 | 1179 | |||
| XM_008760548.1 | XP_008758770.1 | 2 | 506 | |||
| XM_008760549.1 | XP_008758771.1 | 3 | 386 | |||
| XM_008760550.1 | XP_008758772.1 | 2 | 314 | |||
| GenBank mRNA | AF072124.1 | - | 2 | 205 | ||
| BC070936.1 | - | 2 | 205 | 7099111 | ||

Casp7
Genomic Map