siRNA Details
| Gene Symbol: Cabs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022263.2 | NP_071599.2 | 1 | 936 | ||
| GenBank mRNA | AF271155.1 | - | 1 | 632 | ||
| BC079062.1 | - | 1 | 936 | 7115934 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cabs1| Gene Symbol: Cabs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022263.2 | NP_071599.2 | 1 | 936 | ||
| GenBank mRNA | AF271155.1 | - | 1 | 632 | ||
| BC079062.1 | - | 1 | 936 | 7115934 | ||