siRNA Details
| Gene Symbol: Pdcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022265.2 | NP_071601.2 | 8, 9 | 1090 | ||
| XM_006231613.2 | XP_006231675.1 | 9, 10 | 1371 | |||
| XM_006231614.2 | XP_006231676.1 | 9, 10 | 1423 | |||
| XM_006231615.2 | XP_006231677.1 | 9, 10 | 1269 | |||
| XM_006231616.2 | XP_006231678.1 | 9, 10 | 1246 | |||
| GenBank mRNA | AF239739.2 | - | 8, 9 | 1090 | ||
| BC167751.1 | - | 9, 10 | 1278 | 9027822 | ||

Pdcd4
Genomic Map