siRNA Details
| Gene Symbol: Pygl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022268.1 | NP_071604.1 | 9 | 1063 | ||
| GenBank mRNA | BC070901.1 | - | 9 | 1113 | 7100896 | |
| J03080.1 | - | 1 | 52 | |||
| X63515.1 | - | 9 | 1063 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pygl| Gene Symbol: Pygl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022268.1 | NP_071604.1 | 9 | 1063 | ||
| GenBank mRNA | BC070901.1 | - | 9 | 1113 | 7100896 | |
| J03080.1 | - | 1 | 52 | |||
| X63515.1 | - | 9 | 1063 | |||