siRNA Details
| Gene Symbol: Vps33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022286.2 | NP_071622.1 | 22 | 2043 | ||
| XM_008759557.1 | XP_008757779.1 | 19 | 1480 | |||
| GenBank mRNA | BC081707.1 | - | 22 | 2043 | 7128590 | |
| U35245.1 | - | 22 | 1758 | |||

Vps33b
Genomic Map