siRNA Details
| Gene Symbol: Tnmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022290.1 | NP_071626.1 | 4 | 558 | ||
| GenBank mRNA | AB055423.1 | - | 4 | 347 | ||
| AF191769.1 | - | 4 | 558 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnmd| Gene Symbol: Tnmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022290.1 | NP_071626.1 | 4 | 558 | ||
| GenBank mRNA | AB055423.1 | - | 4 | 347 | ||
| AF191769.1 | - | 4 | 558 | |||