siRNA Details
| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022296.2 | NP_071632.2 | 10 | 1971 | ||
| GenBank mRNA | AJ295749.2 | - | 10 | 1970 | ||
| BC105767.1 | - | 10 | 1971 | 7930580 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xylt2| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022296.2 | NP_071632.2 | 10 | 1971 | ||
| GenBank mRNA | AJ295749.2 | - | 10 | 1970 | ||
| BC105767.1 | - | 10 | 1971 | 7930580 | ||