siRNA Details
| Gene Symbol: Scn7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031686.1 | NP_113874.1 | 4 | 550 | ||
| XM_006234296.2 | XP_006234358.1 | 5 | 827 | |||
| XM_006234298.2 | XP_006234360.1 | 5 | 827 | |||
| GenBank mRNA | AJ277392.1 | - | 2 | 145 | ||
| Y09164.1 | - | 4 | 550 | |||

Scn7a
Genomic Map