siRNA Details
| Gene Symbol: Necab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022302.1 | NP_071638.1 | 7 | 668 | ||
| GenBank mRNA | AF193755.1 | - | 7 | 668 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Necab1| Gene Symbol: Necab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022302.1 | NP_071638.1 | 7 | 668 | ||
| GenBank mRNA | AF193755.1 | - | 7 | 668 | ||